Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011249.2 | NP_001011249.1 | 3 - 4 | 280 | 62 | |
XM_012958925.2 | XP_012814379.1 | 5 - 6 | 379 | 62 | ||
XM_012958926.2 | XP_012814380.1 | 4 - 5 | 376 | 62 | ||
XM_012958927.2 | XP_012814381.1 | 5 - 6 | 303 | 62 | ||
XM_012958929.1 | XP_012814383.1 | 5 - 6 | 359 | 62 | ||
XM_012958930.2 | XP_012814384.1 | 5 - 6 | 365 | 62 | ||
XM_012958931.1 | XP_012814385.1 | 4 - 5 | 356 | 62 | ||
XM_012958932.2 | XP_012814386.1 | 4 - 5 | 360 | 62 | ||
GenBank mRNA | BC087828.1 | - | 3 - 4 | 281 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |