Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011255.1 | NP_001011255.1 | 14 - 15 | 1051 | 74 | |
XM_012961026.2 | XP_012816480.1 | 15 - 16 | 1170 | 74 | ||
XM_012961031.2 | XP_012816485.1 | 15 - 16 | 1065 | 74 | ||
GenBank mRNA | BC086500.2 | - | 14 - 15 | 1051 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |