Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011257.1 | NP_001011257.1 | 6 - 7 | 534 | 79 | |
GenBank mRNA | BC087567.2 | - | 6 - 7 | 534 | 79 | |
CR855724.2 | - | 6 - 7 | 505 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |