Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011262.1 | NP_001011262.1 | 5 - 6 | 732 | 63 | |
XM_012961069.2 | XP_012816523.1 | 5 - 6 | 835 | 63 | ||
XM_012961072.2 | XP_012816526.1 | 5 - 6 | 788 | 63 | ||
GenBank mRNA | BC087968.1 | - | 5 - 6 | 732 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |