Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kyat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011283.1 | NP_001011283.1 | 13 - 14 | 1315 | 75 | |
XM_012968302.2 | XP_012823756.1 | 14 - 15 | 1512 | 75 | ||
XM_012968303.2 | XP_012823757.1 | 14 - 15 | 1585 | 75 | ||
XM_012968305.2 | XP_012823759.1 | 14 - 15 | 1414 | 75 | ||
XM_012968307.2 | XP_012823761.1 | 13 - 14 | 1383 | 75 | ||
XM_012968308.2 | XP_012823762.1 | 13 - 14 | 1451 | 75 | ||
XM_012968310.2 | XP_012823764.1 | 12 - 13 | 1322 | 75 | ||
GenBank mRNA | BC087997.1 | - | 13 - 14 | 1315 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |