Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011290.1 | NP_001011290.1 | 6 - 7 | 902 | 102 | |
XM_012964610.2 | XP_012820064.1 | 6 - 7 | 1038 | 102 | ||
GenBank mRNA | BC088007.1 | - | 6 - 7 | 902 | 102 | |
CR848357.2 | - | 6 - 7 | 841 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |