Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203566.1 | NP_988897.1 | 3 - 4 | 399 | 90 | |
GenBank mRNA | BC059734.1 | - | 3 - 4 | 399 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |