Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203570.1 | NP_988901.1 | 3 - 4 | 392 | 73 | |
GenBank mRNA | BC061630.1 | - | 3 - 4 | 392 | 73 | |
BC082501.1 | - | 3 - 4 | 462 | 73 | ||
CR761635.2 | - | 3 - 4 | 391 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |