Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203571.1 | NP_988902.1 | 6 - 7 | 684 | 70 | |
GenBank mRNA | BC061625.1 | - | 6 - 7 | 684 | 70 | |
CR760224.2 | - | 6 - 7 | 689 | 70 | ||
CR760298.2 | - | 2 - 3 | 169 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |