Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203593.1 | NP_988924.1 | 5 - 6 | 481 | 91 | |
GenBank mRNA | BC059752.1 | - | 5 - 6 | 481 | 91 | |
CR760253.2 | - | 5 - 6 | 477 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |