Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203599.1 | NP_988930.1 | 5 - 6 | 798 | 74 | |
XM_012966268.2 | XP_012821722.1 | 5 - 6 | 683 | 74 | ||
GenBank mRNA | BC061431.1 | - | 5 - 6 | 798 | 74 | |
CT025432.1 | - | 5 - 6 | 784 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |