Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011186.2 | NP_001011186.1 | 1 - 2 | 123 | 64 | |
XM_012954851.2 | XP_012810305.1 | 2 - 3 | 627 | 64 | ||
GenBank mRNA | BC086505.2 | - | 1 - 2 | 123 | 64 | |
BC158538.1 | - | 606 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |