Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam241b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012966518.2 | XP_012821972.1 | 2 - 3 | 256 | 68 | |
GenBank mRNA | BC088084.1 | - | 2 - 3 | 161 | 68 | |
CR848180.2 | - | 2 - 3 | 207 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |