Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011331.1 | NP_001011331.1 | 6 - 7 | 993 | 89 | |
XM_012953129.2 | XP_012808583.1 | 6 - 7 | 1123 | 89 | ||
GenBank mRNA | BC088477.1 | - | 6 - 7 | 993 | 89 | |
CR760513.2 | - | 6 - 7 | 996 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |