Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011333.1 | NP_001011333.1 | 5 - 6 | 835 | 94 | |
GenBank mRNA | BC088480.1 | - | 5 - 6 | 835 | 94 | |
CR761106.2 | - | 5 - 6 | 808 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |