Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011373.1 | NP_001011373.1 | 3 - 4 | 835 | 98 | |
GenBank mRNA | BC088548.1 | - | 3 - 4 | 835 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |