Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmynd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011379.1 | NP_001011379.1 | 6 - 7 | 877 | 89 | |
GenBank mRNA | BC088560.1 | - | 6 - 7 | 877 | 89 | |
CR848399.2 | - | 6 - 7 | 880 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |