Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011399.1 | NP_001011399.1 | 8 - 9 | 1140 | 83 | |
XM_018093869.1 | XP_017949358.1 | 8 - 9 | 1100 | 83 | ||
GenBank mRNA | BC088606.1 | - | 8 - 9 | 1140 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |