Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cisd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011479.1 | NP_001011479.1 | 1 - 2 | 125 | 70 | |
XM_012961455.2 | XP_012816909.1 | 1 - 2 | 1114 | 70 | ||
GenBank mRNA | BC088777.1 | - | 1 - 2 | 125 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |