Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tprg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011483.1 | NP_001011483.1 | 3 - 4 | 472 | 96 | |
GenBank mRNA | BC088788.1 | - | 3 - 4 | 472 | 96 | |
CR762132.1 | - | 3 - 4 | 469 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |