Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011491.1 | NP_001011491.1 | 8 - 9 | 1170 | 73 | |
XM_012961018.2 | XP_012816472.1 | 9 - 10 | 1360 | 73 | ||
XM_012961019.2 | XP_012816473.1 | 8 - 9 | 1219 | 73 | ||
XM_012961020.2 | XP_012816474.1 | 7 - 8 | 1106 | 73 | ||
XM_018095376.1 | XP_017950865.1 | 9 - 10 | 1381 | 73 | ||
XM_018095377.1 | XP_017950866.1 | 8 - 9 | 1188 | 73 | ||
GenBank mRNA | BC088876.1 | - | 8 - 9 | 1170 | 73 | |
CR762342.2 | - | 8 - 9 | 1192 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |