Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2d6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015719.1 | NP_001015719.1 | 6 - 7 | 1201 | 66 | |
GenBank mRNA | BC089651.1 | - | 6 - 7 | 1201 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |