Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2d6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015719.1 | NP_001015719.1 | 7 - 8 | 1390 | 83 | |
GenBank mRNA | BC089651.1 | - | 7 - 8 | 1390 | 83 |
Gene Symbol: LOC101731511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004913762.3 | XP_004913819.1 | 7 - 8 | 1198 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |