Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015742.2 | NP_001015742.2 | 3 - 4 | 749 | 105 | |
GenBank mRNA | BC089677.1 | - | 3 - 4 | 723 | 105 | |
BC159065.1 | - | 3 - 4 | 737 | 105 | ||
BC159294.1 | - | 3 - 4 | 728 | 105 | ||
BC170811.1 | - | 3 - 4 | 464 | 105 | ||
BC170813.1 | - | 3 - 4 | 464 | 105 | ||
CR762027.2 | - | 3 - 4 | 649 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |