Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015751.1 | NP_001015751.1 | 3 - 4 | 218 | 104 | |
GenBank mRNA | BC089687.1 | - | 3 - 4 | 218 | 104 | |
CR761507.2 | - | 3 - 4 | 197 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |