Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015761.1 | NP_001015761.1 | 4 - 5 | 597 | 85 | |
XM_012968327.2 | XP_012823781.1 | 5 - 6 | 803 | 85 | ||
XM_012968329.2 | XP_012823783.1 | 5 - 6 | 750 | 85 | ||
GenBank mRNA | BC089697.1 | - | 4 - 5 | 597 | 85 | |
BC121274.1 | - | 3 - 4 | 572 | 85 | ||
BC171186.1 | - | 4 - 5 | 588 | 85 | ||
BC171188.1 | - | 4 - 5 | 588 | 85 | ||
CR848211.2 | - | 4 - 5 | 598 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |