Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015775.1 | NP_001015775.1 | 2 - 3 | 262 | 101 | |
XM_012968332.2 | XP_012823786.1 | 3 - 4 | 464 | 101 | ||
XM_012968333.2 | XP_012823787.1 | 3 - 4 | 349 | 101 | ||
GenBank mRNA | BC089715.1 | - | 2 - 3 | 262 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |