Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015784.1 | NP_001015784.1 | 6 - 7 | 635 | 77 | |
XM_012961652.2 | XP_012817106.1 | 7 - 8 | 853 | 77 | ||
GenBank mRNA | BC089728.1 | - | 6 - 7 | 635 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |