Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015805.2 | NP_001015805.2 | 1 - 2 | 478 | 99 | |
GenBank mRNA | AL848058.2 | - | 1 - 2 | 475 | 96 | |
BC064719.1 | - | 1 - 2 | 411 | 96 | ||
BC090081.1 | - | 1 - 2 | 382 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |