Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015805.2 | NP_001015805.2 | 5 - 6 | 858 | 104 | |
GenBank mRNA | BC064719.1 | - | 5 - 6 | 790 | 104 | |
BC090081.1 | - | 5 - 6 | 762 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |