Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015811.1 | NP_001015811.1 | 184 | 75 | ||
GenBank mRNA | BC090092.1 | - | 184 | 75 | ||
CR762212.2 | - | 98 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |