Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fn3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015844.1 | NP_001015844.1 | 4 - 5 | 580 | 64 | |
GenBank mRNA | BC090133.1 | - | 4 - 5 | 580 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |