Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015845.1 | NP_001015845.1 | 8 - 9 | 907 | 99 | |
GenBank mRNA | BC090134.1 | - | 8 - 9 | 907 | 99 | |
BC122899.1 | - | 8 - 9 | 864 | 99 | ||
CR848552.2 | - | 8 - 9 | 914 | 99 |
Gene Symbol: LOC101731871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004911912.3 | XP_004911969.1 | 8 - 9 | 958 | 99 | |
XM_004911913.3 | XP_004911970.1 | 8 - 9 | 951 | 99 | ||
XM_004911914.3 | XP_004911971.1 | 8 - 9 | 924 | 99 | ||
XM_012957841.2 | XP_012813295.1 | 8 - 9 | 917 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |