Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015899.1 | NP_001015899.1 | 4 - 5 | 518 | 91 | |
XM_012961035.2 | XP_012816489.1 | 3 - 4 | 1771 | 91 | ||
GenBank mRNA | BC118739.1 | - | 4 - 5 | 500 | 91 | |
CR926447.1 | - | 4 - 5 | 518 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |