Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015903.1 | NP_001015903.1 | 2 - 3 | 334 | 93 | |
GenBank mRNA | BC121632.1 | - | 1 - 2 | 164 | 93 | |
BC161440.1 | - | 2 - 3 | 342 | 93 | ||
BC171009.1 | - | 2 - 3 | 290 | 93 | ||
CR926439.1 | - | 2 - 3 | 334 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |