Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015904.2 | NP_001015904.1 | 2 - 3 | 331 | 90 | |
GenBank mRNA | BC075607.1 | - | 2 - 3 | 329 | 90 | |
BC135231.1 | - | 2 - 3 | 345 | 90 | ||
CR926438.2 | - | 2 - 3 | 331 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |