Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016071.2 | NP_001016071.1 | 4 - 5 | 433 | 94 | |
GenBank mRNA | BC170749.1 | - | 3 - 4 | 385 | 94 | |
BC170751.1 | - | 3 - 4 | 385 | 94 | ||
CR762247.2 | - | 4 - 5 | 433 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |