Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016145.2 | NP_001016145.1 | 7 - 8 | 911 | 65 | |
GenBank mRNA | BC091018.1 | - | 7 - 8 | 911 | 65 | |
CR761995.2 | - | 7 - 8 | 824 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |