Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016193.2 | NP_001016193.1 | 6 - 7 | 573 | 88 | |
XM_018095389.1 | XP_017950878.1 | 6 - 7 | 719 | 88 | ||
GenBank mRNA | BC170881.1 | - | 6 - 7 | 573 | 88 | |
BC171266.1 | - | 6 - 7 | 573 | 88 | ||
CR761795.2 | - | 6 - 7 | 573 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |