Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp51a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016194.2 | NP_001016194.1 | 4 - 5 | 842 | 97 | |
GenBank mRNA | BC135319.1 | - | 4 - 5 | 696 | 97 | |
CR926353.2 | - | 4 - 5 | 842 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |