Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016309.2 | NP_001016309.1 | 2 - 3 | 249 | 89 | |
XM_018097103.1 | XP_017952592.1 | 2 - 3 | 183 | 89 | ||
GenBank mRNA | BC091609.1 | - | 2 - 3 | 249 | 89 | |
CR761492.2 | - | 2 - 3 | 139 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |