Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016511.1 | NP_001016511.1 | 2 - 3 | 218 | 79 | |
GenBank mRNA | BC135492.1 | - | 2 - 3 | 186 | 79 | |
BC157293.1 | - | 2 - 3 | 188 | 79 | ||
CR926229.1 | - | 2 - 3 | 218 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |