Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016511.1 | NP_001016511.1 | 4 - 5 | 517 | 83 | |
GenBank mRNA | BC157293.1 | - | 4 - 5 | 487 | 83 | |
CR926229.1 | - | 4 - 5 | 517 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |