Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016558.1 | NP_001016558.1 | 5 - 6 | 738 | 109 | |
XM_012962508.2 | XP_012817962.1 | 6 - 7 | 894 | 109 | ||
GenBank mRNA | CR760702.1 | - | 5 - 6 | 738 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons |