Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016563.1 | NP_001016563.1 | 3 - 4 | 1229 | 69 | |
XM_018093053.1 | XP_017948542.1 | 3 - 4 | 1243 | 69 | ||
GenBank mRNA | CR760685.1 | - | 3 - 4 | 1229 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |