Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gstp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016641.2 | NP_001016641.1 | 2 - 3 | 78 | 64 | |
GenBank mRNA | BC135948.1 | - | 2 - 3 | 82 | 64 | |
CR760401.2 | - | 2 - 3 | 78 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |