Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016644.2 | NP_001016644.1 | 3 - 4 | 300 | 84 | |
GenBank mRNA | CR760398.2 | - | 3 - 4 | 300 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |