Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016647.2 | NP_001016647.1 | 5 - 6 | 1445 | 96 | |
GenBank mRNA | BC135296.1 | - | 5 - 6 | 1499 | 96 | |
CR760380.2 | - | 5 - 6 | 1445 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |