Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016671.2 | NP_001016671.1 | 4 - 5 | 638 | 100 | |
GenBank mRNA | BC157339.1 | - | 4 - 5 | 428 | 100 | |
BC170616.1 | - | 4 - 5 | 609 | 100 | ||
BC170618.1 | - | 4 - 5 | 609 | 100 | ||
CR760242.2 | - | 4 - 5 | 638 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |