Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c11orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016674.2 | NP_001016674.1 | 362 | 60 | ||
GenBank mRNA | BC121515.1 | - | 358 | 60 | ||
CR760234.2 | - | 362 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |